Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178379.3 | NP_848466.1 | 2 | 224 | ||
GenBank mRNA | AK010385.1 | - | 2 | 218 | ||
AK149999.1 | - | 2 | 214 | |||
AK154668.1 | - | 2 | 223 | |||
AK156865.1 | - | 2 | 223 | |||
AK171661.1 | - | 2 | 214 | |||
AK172718.1 | - | 2 | 223 | |||
AK178239.1 | - | 1 | 19 | |||
AK179799.1 | - | 1 | 19 | |||
AK189355.1 | - | 1 | 19 | |||
AK196412.1 | - | 1 | 19 | |||
AK199666.1 | - | 1 | 19 | |||
AK206048.1 | - | 1 | 19 | |||
AK206430.1 | - | 1 | 19 | |||
AK207639.1 | - | 1 | 19 | |||
AK210222.1 | - | 1 | 19 | |||
AK218286.1 | - | 1 | 19 | |||
BC018301.1 | - | 2 | 176 | |||
BC038046.1 | - | 2 | 177 | 4206826 | ||
BY078593.1 | - | 2 | 224 |