Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178379.3 | NP_848466.1 | 2, 3 | 263 | ||
GenBank mRNA | AK010385.1 | - | 2, 3 | 257 | ||
AK149999.1 | - | 2, 3 | 253 | |||
AK154668.1 | - | 2, 3 | 262 | |||
AK156865.1 | - | 2, 3 | 262 | |||
AK171661.1 | - | 2, 3 | 253 | |||
AK172718.1 | - | 2, 3 | 262 | |||
AK178239.1 | - | 1, 2 | 58 | |||
AK179799.1 | - | 1, 2 | 58 | |||
AK189355.1 | - | 1, 2 | 58 | |||
AK196412.1 | - | 1, 2 | 57 | |||
AK199666.1 | - | 1, 2 | 58 | |||
AK206430.1 | - | 1, 2 | 58 | |||
AK207639.1 | - | 1, 2 | 58 | |||
AK210222.1 | - | 1, 2 | 58 | |||
AK212782.1 | - | 1, 2 | 58 | |||
AK212812.1 | - | 1, 2 | 58 | |||
AK218286.1 | - | 1, 2 | 58 | |||
BC018301.1 | - | 2, 3 | 215 | |||
BC038046.1 | - | 2, 3 | 216 | 4206826 | ||
BY078593.1 | - | 2, 3 | 263 |