Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178379.3 | NP_848466.1 | 7 | 1512 | ||
GenBank mRNA | AK010385.1 | - | 7 | 1495 | ||
AK154668.1 | - | 7 | 1511 | |||
AK180475.1 | - | 1 | 83 | |||
AK218809.1 | - | 1 | 78 | |||
AK219091.1 | - | 1 | 78 | |||
BC018301.1 | - | 7 | 1463 | |||
BC038046.1 | - | 7 | 1465 | 4206826 |