Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178392.4 | NP_848479.1 | 9 | 1114 | ||
GenBank mRNA | AK138261.1 | - | 9 | 1115 | ||
AK152833.1 | - | 9 | 1120 | |||
AK207384.1 | - | 2 | 181 | |||
BC030456.1 | - | 9 | 1096 | 5401615 |