Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178599.4 | NP_848714.1 | 5 | 1027 | ||
GenBank mRNA | AK009275.1 | - | 5 | 1016 | ||
AK012653.1 | - | 5 | 1013 | |||
AK013371.1 | - | 5 | 999 | |||
AK017777.1 | - | 6 | 1041 | |||
AK077934.1 | - | 5 | 1027 | |||
AK151560.1 | - | 5 | 1010 | |||
AK158078.1 | - | 6 | 1110 | |||
AK170011.1 | - | 5 | 1000 | |||
BC023888.1 | - | 2 | 699 | |||
BC051517.1 | - | 3 | 787 | |||
BC098183.1 | - | 5 | 1003 | 5150466 |