Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114660.2 | NP_001108132.1 | 4 | 1281 | ||
NM_001286437.1 | NP_001273366.1 | 4 | 1281 | |||
NM_178672.7 | NP_848787.2 | 4 | 1281 | |||
GenBank mRNA | AK034000.1 | - | 4 | 1276 | ||
AK036637.1 | - | 4 | 1223 | |||
AK044408.1 | - | 4 | 1260 | |||
AK047026.1 | - | 4 | 917 | |||
AK047738.1 | - | 4 | 1224 | |||
AK050673.1 | - | 4 | 1232 | |||
AK054548.1 | - | 4 | 1240 | |||
AK088353.1 | - | 4 | 1167 | |||
AK089011.1 | - | 4 | 982 | |||
AK135666.1 | - | 4 | 517 | |||
AK140246.1 | - | 4 | 1282 | |||
AK143615.1 | - | 4 | 1261 | |||
AK163028.1 | - | 4 | 1240 | |||
BC042528.1 | - | 4 | 1292 | 3256631 |