Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178675.4 | NP_848790.2 | 8 | 2504 | ||
GenBank mRNA | AK038601.1 | - | 8 | 2499 | ||
AK048957.1 | - | 8 | 2464 | |||
AK053426.1 | - | 8 | 2473 | |||
AK053545.1 | - | 8 | 2443 | |||
AK083768.1 | - | 8 | 2443 | |||
AK139331.1 | - | 8 | 2474 | |||
AK163806.1 | - | 8 | 2304 | |||
AK165021.1 | - | 8 | 2458 | |||
AK166268.1 | - | 8 | 2436 | |||
BC059075.1 | - | 8 | 2400 | 6826829 |