Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145454.2 | NP_001138926.1 | 4 | 523 | ||
NM_005563.3 | NP_005554.1 | 4 | 523 | |||
NM_203399.1 | NP_981944.1 | 4 | 499 | |||
NM_203401.1 | NP_981946.1 | 4 | 711 | |||
GenBank mRNA | AB209282.1 | - | 3 | 2385 | ||
AB451319.1 | - | 2 | 347 | |||
AB451448.1 | - | 2 | 347 | |||
AK054594.1 | - | 5 | 419 | |||
AK303692.1 | - | 4 | 439 | |||
AK311801.1 | - | 4 | 440 | |||
BC014353.1 | - | 4 | 492 | 3683516 | ||
BC082228.1 | - | 3 | 421 | 2822803 | ||
BG030679.1 | - | 4 | 494 | |||
BG493669.1 | - | 4 | 525 | |||
BM563894.1 | - | 4 | 544 | |||
BQ428400.1 | - | 4 | 744 | |||
BX647885.1 | - | 3 | 2399 | |||
J04991.1 | - | 4 | 450 | |||
X53305.1 | - | 4 | 428 | |||
Z11566.1 | - | 4 | 438 |