Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181072.3 | NP_851417.2 | 8, 9 | 1139 | ||
GenBank mRNA | AK029145.1 | - | 8, 9 | 1141 | ||
BC040786.1 | - | 9, 10 | 1195 | |||
BC051391.1 | - | 8, 9 | 1121 | 5053090 | ||
CX242330.1 | - | 3, 4 | 337 |