Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005567.3 | NP_005558.1 | 5, 6 | 816 | ||
GenBank mRNA | AK055977.1 | - | 4, 5 | 479 | ||
AK057776.1 | - | 4, 5 | 447 | |||
AK293183.1 | - | 4, 5 | 490 | |||
AK301040.1 | - | 4, 5 | 724 | |||
AK301444.1 | - | 4, 5 | 464 | |||
AK307665.1 | - | 5, 6 | 737 | |||
BC002403.2 | - | 5, 6 | 749 | 2822371 | ||
BC002998.1 | - | 5, 6 | 749 | 2822371 | ||
BC015761.1 | - | 5, 6 | 781 | 4858122 | ||
KU178164.1 | - | 4, 5 | 637 | |||
L13210.1 | - | 5, 6 | 816 | |||
X79089.1 | - | 5, 6 | 768 |