Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183034.1 | NP_898855.1 | 4 | 502 | ||
GenBank mRNA | AK154703.1 | - | 3 | 431 | ||
AK155357.1 | - | 4 | 526 | |||
AK157752.1 | - | 4 | 537 | |||
AK190944.1 | - | 1 | 26 | |||
BC053079.1 | - | 4 | 502 | 5702529 |