Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005575.2 | NP_005566.2 | 2 | 714 | ||
NM_175920.3 | NP_787116.2 | 2 | 506 | |||
GenBank mRNA | AK314226.1 | - | 2 | 256 | ||
D50810.1 | - | 2 | 671 | |||
U62768.1 | - | 2 | 261 | |||
U62769.1 | - | 2 | 516 |