Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183193.2 | NP_899016.1 | 2 | 2055 | ||
GenBank mRNA | AK045276.1 | - | 2 | 2055 | ||
BC096623.1 | - | 2 | 2124 | 30434074 |