Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marveld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183195.2 | NP_899018.1 | 1 | 1902 | ||
GenBank mRNA | AK081212.1 | - | 1 | 1585 | ||
AK132243.1 | - | 1 | 1893 | |||
BC044794.1 | - | 1 | 328 | |||
BC054384.1 | - | 1 | 1865 | 6475446 |