Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB21L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005584.4 | NP_005575.1 | 1 | 1177 | ||
GenBank mRNA | AB073388.1 | - | 1 | 1167 | ||
AB593021.1 | - | 1 | 1098 | |||
AB593022.1 | - | 1 | 1168 | |||
AK313185.1 | - | 1 | 1153 | |||
BC028170.1 | - | 1 | 1098 | 5244028 | ||
CR457420.1 | - | 1 | 620 |