Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163486.1 | NP_001156958.1 | 6 | 812 | ||
NM_198030.2 | NP_932147.2 | 6 | 812 | |||
GenBank mRNA | AY101768.1 | - | 6 | 739 | ||
BC019427.1 | - | 6 | 788 | 5132342 | ||
DV660487.1 | - | 3 | 238 |