Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198101.1 | NP_932769.1 | 10 | 1002 | ||
GenBank mRNA | AK052221.1 | - | 10 | 967 | ||
AK082337.1 | - | 10 | 999 | |||
AK137177.1 | - | 8 | 995 | |||
AK172438.1 | - | 10 | 990 | |||
BC057037.1 | - | 10 | 1002 | 5702855 | ||
BC060628.1 | - | 10 | 977 | 6847400 |