Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159288.1 | NP_001152760.1 | 18, 19 | 2455 | ||
NM_198162.1 | NP_937805.1 | 17, 18 | 1782 | |||
GenBank mRNA | AK173042.1 | - | 18, 19 | 2455 | ||
BC059243.1 | - | 17, 18 | 1782 | 6417551 | ||
BC063082.1 | - | 6, 7 | 722 |