Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145921.1 | NP_001139393.1 | 11 | 2429 | ||
NM_001145922.1 | NP_001139394.1 | 11 | 2429 | |||
NM_198249.4 | NP_937892.2 | 11 | 2429 | |||
GenBank mRNA | AK131136.1 | - | 11 | 2398 | ||
AK143527.1 | - | 11 | 2404 | |||
AK147761.1 | - | 11 | 2365 | |||
AK163945.1 | - | 11 | 2430 | |||
BC059018.1 | - | 11 | 2331 | 6848691 |