Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198250.1 | NP_937893.1 | 3 | 1780 | ||
GenBank mRNA | AK043555.1 | - | 1 | 570 | ||
AK140418.1 | - | 2 | 1727 | |||
BC053486.1 | - | 2 | 1644 | |||
BC060263.1 | - | 3 | 1780 | 5686281 |