Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198610.2 | NP_941012.1 | 3 | 723 | ||
GenBank mRNA | AK137439.1 | - | 3 | 717 | ||
BC055811.1 | - | 3 | 696 | 6414125 |