Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168624.1 | NP_001162095.1 | 2 | 481 | ||
NM_198614.3 | NP_941016.2 | 2 | 607 | |||
GenBank mRNA | AK029946.1 | - | 2 | 482 | ||
BC089008.1 | - | 2 | 589 | 30537363 |