Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168624.1 | NP_001162095.1 | 2 | 2219 | ||
NM_198614.3 | NP_941016.2 | 2 | 2345 | |||
GenBank mRNA | AK029946.1 | - | 2 | 2220 | ||
BC089008.1 | - | 2 | 2328 | 30537363 |