Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142804.1 | NP_001136276.1 | 1 | 320 | ||
NM_198636.3 | NP_941038.2 | 1 | 320 | |||
GenBank mRNA | AK033376.1 | - | 1 | 325 | ||
AK163897.1 | - | 1 | 307 | |||
BC113198.1 | - | 1 | 284 | 40062399 | ||
BY720548.1 | - | 1 | 320 |