Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142804.1 | NP_001136276.1 | 2 | 498 | ||
NM_198636.3 | NP_941038.2 | 2 | 498 | |||
GenBank mRNA | AK033376.1 | - | 2 | 503 | ||
BC113198.1 | - | 2 | 462 | 40062399 | ||
BY720548.1 | - | 2 | 498 |