Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105243590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB090859.1 | - | 1 | 127 | ||
AF023204.1 | - | 1 | 108 | |||
AF163758.1 | - | 1 | 127 | |||
AF165119.1 | - | 1 | 100 | |||
AF303836.1 | - | 1 | 127 | |||
AF547084.1 | - | 1 | 103 | |||
AY174016.1 | - | 1 | 103 | |||
AY369973.1 | - | 1 | 103 | |||
FJ816503.1 | - | 52 | ||||
M54977.1 | - | 220 |
Gene Symbol: Igh-VJ558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF296418.1 | - | 1 | 127 | ||
AF296419.1 | - | 1 | 127 | |||
AF296420.1 | - | 1 | 127 | |||
U40318.1 | - | 1 | 109 | |||
U58290.1 | - | 1 | 109 |
Gene Symbol: Igha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136471.1 | - | 239 |
Gene Symbol: Ighg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC055910.1 | - | 221 |
Gene Symbol: Ighv1-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF296421.1 | - | 1 | 127 |
Gene Symbol: Ighv1-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U41426.1 | - | 1 | 127 |
Gene Symbol: Ighv1-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ340232.1 | - | 103 | |||
M31276.1 | - | 130 | ||||
M31277.1 | - | 130 |