Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5031414D18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198642.2 | NP_941044.3 | 2 | 342 | ||
GenBank mRNA | AK155201.1 | - | 2 | 328 | ||
AK170426.1 | - | 2 | 306 | |||
BC052668.1 | - | 2 | 354 | 30054870 |