Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6430706D22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032612.1 | - | 2 | 1599 | ||
AK135880.1 | - | 1 | 1120 | |||
BC062125.1 | - | 5 | 2007 | 6826662 |
Gene Symbol: Hjurp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198652.2 | NP_941054.2 | 8 | 2546 | ||
GenBank mRNA | BC057309.1 | - | 8 | 2487 | 6826508 |