Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198653.2 | NP_941055.1 | 3 | 560 | ||
GenBank mRNA | AK036182.1 | - | 3 | 528 | ||
AK043590.1 | - | 3 | 562 | |||
AK043729.1 | - | 3 | 538 | |||
AK048653.1 | - | 3 | 561 | |||
AK050358.1 | - | 3 | 543 | |||
AK051778.1 | - | 3 | 521 | |||
AK078183.1 | - | 3 | 561 | |||
AK085714.1 | - | 3 | 557 | |||
AK088965.1 | - | 3 | 521 | |||
AK164435.1 | - | 3 | 512 | |||
AK166832.1 | - | 3 | 543 | |||
BC052403.1 | - | 3 | 537 | 5715723 |