Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198653.2 | NP_941055.1 | 10, 11 | 1437 | ||
GenBank mRNA | AK036182.1 | - | 10, 11 | 1405 | ||
AK043590.1 | - | 10, 11 | 1439 | |||
AK043729.1 | - | 10, 11 | 1415 | |||
AK048653.1 | - | 10, 11 | 1438 | |||
AK050358.1 | - | 10, 11 | 1420 | |||
AK051778.1 | - | 10, 11 | 1398 | |||
AK078183.1 | - | 10, 11 | 1439 | |||
AK085714.1 | - | 10, 11 | 1434 | |||
AK088965.1 | - | 10, 11 | 1398 | |||
AK166832.1 | - | 10, 11 | 1420 | |||
BC037123.1 | - | 5, 6 | 497 | |||
BC052403.1 | - | 10, 11 | 1414 | 5715723 |