Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9430007A20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198662.3 | NP_941064.2 | 2 | 342 | ||
GenBank mRNA | AK034567.1 | - | 2 | 343 | ||
BC148288.1 | - | 2 | 311 |