Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199301.2 | NP_955005.2 | 1 | 176 | ||
GenBank mRNA | AK140018.1 | - | 1 | 92 | ||
AK144453.1 | - | 1 | 101 | |||
AK144625.1 | - | 1 | 134 | |||
BC027306.1 | - | 1 | 85 | 4019717 | ||
BY240360.1 | - | 1 | 176 |