Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114361.1 | NP_001107833.1 | 21 | 2527 | ||
NM_001114362.1 | NP_001107834.1 | 19 | 2320 | |||
NM_001286567.1 | NP_001273496.1 | 21 | 2430 | |||
NM_199466.3 | NP_955760.3 | 18 | 2191 | |||
GenBank mRNA | AK019611.1 | - | 2 | 149 | ||
AK077527.1 | - | 13 | 1356 | |||
AK139067.1 | - | 5 | 384 | |||
AK144604.1 | - | 21 | 2528 | |||
AK189201.1 | - | 4 | 279 | |||
BC032258.1 | - | 7 | 675 | |||
BC065178.1 | - | 21 | 2368 | 6827397 | ||
BC066099.1 | - | 18 | 2122 | 30533573 | ||
BC067011.1 | - | 18 | 2140 | 6405184 | ||
BC070427.1 | - | 19 | 2260 | 6413041 | ||
BC079619.1 | - | 18 | 2122 | 30533573 | ||
DQ230326.1 | - | 20 | 2210 |