Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291424.1 | NP_001278353.1 | 7 | 3284 | ||
NM_005475.2 | NP_005466.1 | 8 | 4207 | |||
XM_005253818.4 | XP_005253875.1 | 9 | 4351 | |||
XM_005253819.4 | XP_005253876.1 | 8 | 4234 | |||
XM_006719180.3 | XP_006719243.1 | 7 | 3836 | |||
XM_011537719.2 | XP_011536021.1 | 9 | 4354 | |||
XM_011537720.2 | XP_011536022.1 | 9 | 4019 | |||
XM_011537721.2 | XP_011536023.1 | 7 | 5819 | |||
GenBank mRNA | AB208911.1 | - | 7 | 3444 | ||
AF055581.1 | - | 8 | 4205 | |||
AK025666.1 | - | 1 | 1214 | |||
AK025745.1 | - | 1 | 2051 | |||
BC015786.1 | - | 1 | 544 |