Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199473.2 | NP_955767.1 | 4 | 4192 | ||
GenBank mRNA | AK076518.1 | - | 1475 | |||
AK131153.1 | - | 1 | 3817 | |||
BC065148.1 | - | 4 | 4191 | 30355747 | ||
BC080317.1 | - | 5 | 4012 | 6853989 |