Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005481.2 | NP_005472.2 | 11 | 1965 | ||
XM_017026120.1 | XP_016881609.1 | 10 | 1753 | |||
XM_017026121.1 | XP_016881610.1 | 10 | 1805 | |||
GenBank mRNA | AF106934.1 | - | 11 | 1952 | ||
AF121228.1 | - | 10 | 1814 | |||
AL137732.1 | - | 4 | 667 | |||
BC004554.1 | - | 11 | 1946 | 4053743 | ||
BC007853.2 | - | 7 | 1075 | |||
BC011841.1 | - | 11 | 1946 | 4053664 | ||
BC017282.2 | - | 11 | 1940 | 4876977 | ||
EU392448.1 | - | 1 | 20 | |||
EU392450.1 | - | 1 | 20 | |||
EU392458.1 | - | 4 | 424 | |||
EU392459.1 | - | 4 | 415 | |||
EU392497.1 | - | 1 | 20 |