Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201370.2 | NP_958758.2 | 12 | 2269 | ||
GenBank mRNA | AK139573.1 | - | 12 | 2228 | ||
AK143336.1 | - | 12 | 2193 | |||
AK163344.1 | - | 12 | 2270 | |||
BC052883.1 | - | 12 | 2176 | 30053490 | ||
BC080784.1 | - | 12 | 2194 | 30481521 | ||
DQ011691.1 | - | 12 | 2090 |