Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201406.1 | NP_958808.1 | 11, 12 | 1453 | ||
GenBank mRNA | AK033331.1 | - | 11, 12 | 1427 | ||
AK039604.1 | - | 11, 12 | 1424 | |||
AK043232.1 | - | 11, 12 | 1427 | |||
AK045759.1 | - | 11, 12 | 1423 | |||
AK053771.1 | - | 11, 12 | 1427 | |||
AK142871.1 | - | 11, 12 | 1424 | |||
AK151858.1 | - | 11, 12 | 1415 | |||
AK152229.1 | - | 11, 12 | 1415 | |||
AK153300.1 | - | 11, 12 | 1419 | |||
AK166408.1 | - | 11, 12 | 1431 | |||
AK169633.1 | - | 10, 11 | 1226 | |||
AK216113.1 | - | 2, 3 | 316 | |||
BC058979.1 | - | 11, 12 | 1453 | 5716747 |