Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201407.4 | NP_958809.3 | 11 | 1741 | ||
GenBank mRNA | AK155638.1 | - | 11 | 1723 | ||
AY512564.1 | - | 9 | 1346 | |||
BC049972.1 | - | 1 | 27 | |||
BC066102.1 | - | 10 | 1520 | 30531856 | ||
BC151194.1 | - | 10 | 1387 | 9088082 |