Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039514.1 | NP_001034603.1 | 2 | 442 | ||
GenBank mRNA | AK159104.1 | - | 2 | 443 | ||
AK177102.1 | - | 1 | 16 | |||
AK177852.1 | - | 1 | 16 | |||
AK185697.1 | - | 1 | 16 | |||
AK213599.1 | - | 1 | 16 | |||
BC061234.1 | - | 2 | 403 |