Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201411.2 | NP_958813.1 | 2 | 1057 | ||
GenBank mRNA | AK031503.1 | - | 2 | 1057 | ||
AY495668.1 | - | 2 | 1012 | |||
BC070403.1 | - | 2 | 1061 | 5696478 | ||
BC111879.1 | - | 2 | 760 | 40059728 | ||
BC112383.1 | - | 2 | 760 | 40059727 | ||
BC138214.1 | - | 2 | 698 | 8861233 |