Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201531.3 | NP_963289.2 | 1 | 936 | ||
GenBank mRNA | AK137367.1 | - | 1 | 629 | ||
BC053089.1 | - | 1 | 573 | 5709192 |