Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135729.1 | NP_001129201.1 | 15 | 1509 | ||
NM_001135730.1 | NP_001129202.1 | 14 | 1444 | |||
NM_001135732.1 | NP_001129204.1 | 15 | 1582 | |||
NM_005488.2 | NP_005479.1 | 15 | 1579 | |||
NR_024194.1 | 15 | 1608 | ||||
NR_024195.1 | 14 | 1529 | ||||
XM_011529818.2 | XP_011528120.1 | 16 | 1596 | |||
XM_011529820.2 | XP_011528122.1 | 14 | 1447 | |||
XM_017028529.1 | XP_016884018.1 | 16 | 1593 | |||
XM_017028530.1 | XP_016884019.1 | 14 | 1444 | |||
GenBank mRNA | AF040251.1 | - | 1 | 103 | ||
AJ006973.1 | - | 15 | 1515 | |||
AK026576.1 | - | 15 | 1532 | |||
AK090723.1 | - | 3 | 1088 | |||
AK091958.1 | - | 3 | 1770 | |||
AK097066.1 | - | 13 | 1090 | |||
AK097926.1 | - | 17 | 1795 | |||
AK293694.1 | - | 15 | 1509 | |||
AK295918.1 | - | 14 | 1439 | |||
AK296670.1 | - | 15 | 1516 | |||
AK297827.1 | - | 14 | 1358 | |||
AK314522.1 | - | 15 | 1488 | |||
BC040006.1 | - | 15 | 1472 | |||
BC046151.1 | - | 15 | 1460 | 6470390 | ||
CR456595.1 | - | 15 | 1475 |