Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015880.1 | NP_001015880.1 | 5 | 800 | ||
NM_004670.3 | NP_004661.2 | 5 | 800 | |||
GenBank mRNA | AF074331.1 | - | 5 | 600 | ||
AF091242.1 | - | 5 | 617 | |||
AF150754.2 | - | 5 | 617 | |||
AF173365.1 | - | 5 | 585 | |||
AF313907.1 | - | 5 | 537 | |||
AK301453.1 | - | 5 | 587 | |||
AK309262.1 | - | 5 | 609 | |||
AK312556.1 | - | 5 | 641 | |||
BC009894.2 | - | 5 | 648 | 2988345 | ||
CB960618.1 | - | 5 | 679 |