Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008551.1 | NP_032577.1 | 7 | 848 | ||
GenBank mRNA | BC024559.1 | - | 3 | 217 | ||
BC063064.1 | - | 7 | 848 | 5687007 | ||
X76850.1 | - | 7 | 827 |