Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042743.2 | NP_001036208.1 | 2 | 541 | ||
NM_008641.3 | NP_032667.2 | 2 | 541 | |||
GenBank mRNA | AK086418.1 | - | 1 | 845 | ||
BC060703.1 | - | 2 | 111 | 6412811 | ||
CB246117.1 | - | 2 | 282 | |||
U02313.1 | - | 2 | 208 |