Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042743.2 | NP_001036208.1 | 11 | 1358 | ||
NM_008641.3 | NP_032667.2 | 10 | 1337 | |||
GenBank mRNA | BC060703.1 | - | 11 | 928 | 6412811 | |
BU055783.1 | - | 8 | 634 | |||
BU701350.1 | - | 1 | 50 | |||
U02313.1 | - | 10 | 1007 |