Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043942.1 | - | 2 | 2070 | ||
AK047847.1 | - | 3 | 2102 | |||
AK163438.1 | - | 3 | 1892 | |||
BC068137.1 | - | 2 | 2208 | |||
BC079608.1 | - | 2 | 2158 |