Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005493.2 | NP_005484.2 | 14 | 2593 | ||
XM_006714945.2 | XP_006715008.1 | 14 | 2073 | |||
XM_011514205.2 | XP_011512507.1 | 12 | 2341 | |||
XM_017010149.1 | XP_016865638.1 | 14 | 1981 | |||
GenBank mRNA | AB008515.1 | - | 14 | 2282 | ||
AB055311.1 | - | 14 | 2593 | |||
AF306510.1 | - | 14 | 2131 | |||
BC052781.1 | - | 14 | 2754 | |||
BC063849.1 | - | 12 | 1849 | 6186252 |