Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008957.2 | NP_032983.1 | 8 | 1051 | ||
GenBank mRNA | AK147626.1 | - | 8 | 1260 | ||
U46155.1 | - | 8 | 1051 |